Order Entry
Czech Republic
ContactUsLinkComponent
Phenolphthalein ≥98%, white powder USP, NF
Health
Phenolphthalein ≥98%, white powder USP, NF
Supplier:  MP Biomedicals
Certificates
undefined
Health
Phenolphthalein ≥98%, white powder USP, NF
Supplier:  MP Biomedicals

Specification Test Results

  • Assay:
    ≥98%
  • Colour of solution:
    Passes
  • Identification:
    Passes
  • Melting Temperature:
    ≥258 °C
  • Loss on Drying:
    ≤1.0%
  • Residue on Ignition:
    ≤0.1%
  • Heavy Metals:
    ≤0.0015%
  • Limit of Fluoran:
    Passes
  • Chromatographic Purity:
    Passes
  • Organic Volatile Impurities:
    Passes

Specification Test Results

Frequently Bought Together