
Phenolphthalein ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-09-8
:
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913
:318.33 g/mol
Merck Index:14,7243
Melting Pt:258…262 °C
Density:1.299
EINECS:201-004-7
REACH:01-2119498295-24
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together