Order Entry
Czech Republic
ContactUsLinkComponent
4-Hydroxybenzophenone for synthesis, Sigma-Aldrich®
4-Hydroxybenzophenone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
4-Hydroxybenzophenone for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    131 - 133 °C
  • Molar Mass:
    198.22 g/mol
  • Boiling Point:
    260 - 262 °C (32 hPa)
  • Synonyms:
    p-Benzoylphenol
  • Hill Formula:
    C13H10O2
  • Chemical Formula:
    4-(OH)C6H4COC6H5

Specification Test Results

Related Information

Frequently Bought Together