
4-Aminobenzophenone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1137-41-3
:
Formula:C₁₃H₁₁NO
MDL Number:MFCD00007895
:197.24 g/mol
Boiling Pt:246 °C (17hPa)
Melting Pt:120…124 °C
Storage Temperature:Ambient
EINECS:214-506-6
:GHS07
Specification Test Results
- Appearance:Yellow to yellow-green to beige to light grey to orange to brown Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:120 to 124 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together