Order Entry
Czech Republic
ContactUsLinkComponent
 
Bromophenol blue, pure indicator
 
undefined
Bromophenol blue, pure indicator
 

 

  • Appearance:
    Pink-orange to red Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 % in ethanol) Clear
  • Specific extinction E (1%/1cm):
    ≥1100 (592 nm, in buffer)
  • Visual transition interval:
    from pH 3.0 (yellow) to pH 4.6 (blue)

 

 

Frequently Bought Together