VWR® PerfectBlue Mini M, Gel System
700-0758
- Description:PerfectBlue gel system Mini M
- Seq:01
- Model:Mini M
- Buffer volume:600 ml ml
- Run time:≥45 h
- Gel size W×L:90×110 mm
- Unit W×L×H:90×110×95 mm
- Pk:1
The PerfectBlue Mini M electrophoresis system is a very convenient size for a wide range of routine uses. Supplied with two combs, but with a wide range of standard and microtitre combs available, this system can be used for the separation of up to 36 samples. The system comes 'ready to use' with lid, power leads and a gel tray. Its high specification construction and embedded platinum electrodes provide high quality results time after time.
*Based on an agarose gel of approximately 5 mm thickness
: System is supplied with 2 combs (10 and 14 teeth, 1,5 mm thickness) as standard.