Order Entry
Czech Republic
ContactUsLinkComponent
 
VWR® PerfectBlue Mini S, Gel System
Catalog # 700-0741
Supplier:  VWR Collection
undefined
VWR® PerfectBlue Mini S, Gel System
Catalog # 700-0741
Supplier:  VWR Collection

Specifications

  • Description:
    PerfectBlue gel system Mini S
  • Seq:
    01
  • Model:
    Mini S
  • Gel volume:
    ~30 ml
  • Buffer volume:
    ~400 ml
  • Run time:
    ≥30 h
  • Gel size W×L:
    70×80 mm
  • Unit W×L×H:
    120×160×95 mm
  • Pk:
    1

Specifications

About this item

The smallest of all the PerfectBlue electrophoresis systems, the Mini S can be used for up to 24 samples enabling short and rapid sample separation. This model includes all the features of the larger models, including direct chamber gel casting and embedded platinum electrodes, and is supplied ready to use with lid, power leads, two combs and a gel tray with an easy to read ruler.

Gel size (W×L): 70×80 mm

Accessories information: * Based on an agarose gel of approximately 5 mm thickness
Ordering information: System is supplied with 2 combs (6 and 10 teeth, 1,5 mm thickness) as standard.

Combs

Product Information