sec-Pentylbenzene tech. 85%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2719-52-0
:
Formula:C₆H₅CH(CH₃)(CH₂)₂CH₃
MDL Number:MFCD00027145
:148.25 g/mol
Boiling Pt:199 °C (1013 hPa)
Melting Pt:–48 °C
Density:0.86
Storage Temperature:Ambient
EINECS:220-324-8
Related Information
Frequently Bought Together