ContactUsLinkComponent
ESD safety shoes, lace-up, uvex 1 x-tended support, 8512
ESD safety shoes, lace-up, uvex 1 x-tended support, 8512
Catalog # UVEX8512.7/43
Supplier:  UVEX
undefined
ESD safety shoes, lace-up, uvex 1 x-tended support, 8512
Catalog # UVEX8512.7/43
Supplier:  UVEX
Specifications
  • Shoe size:
    43
  • Colour:
    Black/red
  • Pk:
    1 PAIR
Specifications
About this item
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Side frame offers medial and lateral support to protect against ankle injuries
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • Elastic laces for quick and individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black/red
Width: 10 or 11
Certifications: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC