
p-Toluenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:98-59-9
:
Formula:C₇H₇ClO₂S
MDL Number:MFCD00007450
:190.65 g/mol
Merck Index:13,09612
Boiling Pt:265 °C (1013 hPa)
Melting Pt:67 °C
Density:1.49 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-684-8
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ200
Flash Pt:128 °C (closed cup)
:GHS05
Frequently Bought Together