Order Entry
Czech Republic
ContactUsLinkComponent
4-Toluenethiol for synthesis, Sigma-Aldrich®
Irritant
4-Toluenethiol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
4-Toluenethiol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    39 - 42 °C
  • Molar Mass:
    124.21 g/mol
  • Boiling Point:
    195 °C (1013 hPa)
  • Flash Point:
    70 °C

Specification Test Results

Related Information

Frequently Bought Together