

4-Toluenethiol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-45-6
:
Formula:C₇H₈S
MDL Number:MFCD00004851
:124.21 g/mol
Merck Index:14,9325
Boiling Pt:195 °C
Melting Pt:40…44 °C
Density:1.022 g/cm³ (51 °C)
Flash Pt:68 °C (154 °F)
Storage Temperature:Ambient
EINECS:203-399-1
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together