Order Entry
Czech Republic
ContactUsLinkComponent
o-Cresol, Supelco®
Toxic
Corrosive
o-Cresol, Supelco®
Catalog # 36922-250MG
Supplier:  Merck
CAS Number:  
undefined
Toxic
Corrosive
o-Cresol, Supelco®
Catalog # 36922-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :108.14 g/mol
Merck Index:13,02604
Boiling Pt:191 °C (1013 hPa)
Melting Pt:30…34 °C
Density:1.047 g/cm³ (20 °C)
Flash Pt:81 °C
Storage Temperature:Ambient
EINECS:202-423-8
UN:3455
ADR:6.1,II
 :GHS06
 :GHS05
CAS Number:95-48-7
Formula:C₇H₈O
MDL Number:MFCD00002226