Order Entry
Czech Republic
ContactUsLinkComponent
Benzyl alcohol 98.0-100.5% NF, Macron Fine Chemicals™
Benzyl alcohol 98.0-100.5% NF, Macron Fine Chemicals™
Certificates
undefined
Benzyl alcohol 98.0-100.5% NF, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    98.0 - 100.5 %
  • NF - Assay (C₆H₅CH₂OH):
    98.0 - 100.5 %
  • NF - Acidity:
    Passes Test
  • NF - Clarity of Solution:
    Passes Test
  • NF - Color of Solution:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Residue on Evaporation:
    ≤0.05 %
  • NF - Peroxide Value:
    ≤5
  • NF - Refractive Index, n²ºD:
    1.538 - 1.541
  • NF - Benzaldehyde and Other Related Substances - Benzaldehyde:
    ≤0.15 %
  • NF - Benzaldehyde and Other Related Substances - Cyclohexylmethanol:
    ≤0.10 %
  • NF - Benzaldehyde and Other Related Substances - Total Other Peaks Less:
    ≤0.04 %
  • Than RT of Benzyl Alcohol:
    ≤0.04 %
  • NF - Benzaldehyde and Other Related Substances - Total Other Peaks:
    ≤0.3 %
  • Greater Than RT of Benzyl Alcohol:
    ≤0.3 %
  • NF - Benzene:
    ≤2 ppm
  • Solubility:
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together