
2-Methyl-1-phenyl-2-propanol, Sigma-Aldrich®
Catalog # SIAL170275-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:150.22 g/mol
Boiling Pt:215 °C (1013 hPa)
Melting Pt:23…25 °C
Density:0.974 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:202-896-0
:GHS07
CAS Number:100-86-7
Formula:C₆H₅CH₂C(CH₃)(OH)CH₃
MDL Number:MFCD00004465