
2-Phenyl-2-propanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:617-94-7
:
Formula:C₆H₅C(OH)(CH₃)₂
MDL Number:MFCD00004456
:136.19 g/mol
Boiling Pt:215 °C (1013 hPa)
Melting Pt:33…35 °C
Density:0.973 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:210-539-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together