
3,5-Dimethoxytoluene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4179-19-5
:
Formula:C₉H₁₂O₂
MDL Number:MFCD00015435
:152.19 g/mol
Boiling Pt:244 °C
Density:1.039
Flash Pt:101 °C (213 °F)
EINECS:224-048-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together