Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Hydroxy-4-methoxybenzyl alcohol
3-Hydroxy-4-methoxybenzyl alcohol, Sigma-Aldrich®
3-Hydroxy-4-methoxybenzyl alcohol, Sigma-Aldrich®
3-Hydroxy-4-methoxybenzyl alcohol, Sigma-Aldrich®
SIAL188433-10G
:
Merck
:
3-Hydroxy-4-methoxybenzyl alcohol, Sigma-Aldrich®
SIAL188433-10G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
154.17 g/mol
:
M-Clarity™ quality level = MQ100
:
224-489-7
:
4383-06-6
:
C₈H₁₀O₃
:
MFCD00004644