Order Entry
Czech Republic
ContactUsLinkComponent
2-Methoxybenzyl alcohol, Sigma-Aldrich®
2-Methoxybenzyl alcohol, Sigma-Aldrich®
Catalog # SIALM10808-5G
Supplier:  Merck
CAS Number:  
2-Methoxybenzyl alcohol, Sigma-Aldrich®
Catalog # SIALM10808-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :138.17 g/mol
Boiling Pt:250 °C (1013 hPa)
Density:1.12 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:210-296-5
CAS Number:612-16-8
Formula:C₈H₁₀O₂
MDL Number:MFCD00004611