
6-Nitro-m-cresol, Sigma-Aldrich®
Catalog # SIAL137790-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:153.14 g/mol
Melting Pt:53…56 °C
Storage Temperature:Ambient
UN:2446
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:211-843-0
:GHS05
CAS Number:700-38-9
Formula:C₇H₇NO₃
MDL Number:MFCD00007111