Zadání objednávky
Czech Republic
ContactUsLinkComponent
6-Nitro-m-cresol 97%
Irritant
6-Nitro-m-cresol 97%
Certificates
undefined
Irritant
6-Nitro-m-cresol 97%

Specification Test Results

  • Appearance:
    Yellow to yellow-green Crystalline powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    53 to 56 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together