Order Entry
Czech Republic
ContactUsLinkComponent
4-Nitro-m-cresol
Irritant
4-Nitro-m-cresol
Catalog # EHERC15110000
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
4-Nitro-m-cresol
Catalog # EHERC15110000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
Melting Pt:126 °C
Storage Temperature:Ambient
EINECS:219-952-5
ADR:6.1,III
 :153.14 g/mol
UN:2446
 :GHS07
CAS Number:2581-34-2
Formula:C₇H₇NO₃
MDL Number:MFCD00007333