3-Nitro-o-cresol, Sigma-Aldrich®
Catalog # SIAL106941-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:153.14 g/mol
Melting Pt:149…152 °C
UN:2446
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:226-739-0
CAS Number:5460-31-1
Formula:C₇H₇NO₃
MDL Number:MFCD00007241