Order Entry
Czech Republic
ContactUsLinkComponent
3-Nitro-o-cresol
3-Nitro-o-cresol
Catalog # EHERC15108500
Supplier:  EHRENSTORFER
CAS Number:  
3-Nitro-o-cresol
Catalog # EHERC15108500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :153.14 g/mol
Melting Pt:149…152 °C
EINECS:226-739-0
UN:2446
ADR:6.1,III
CAS Number:5460-31-1
Formula:C₇H₇NO₃
MDL Number:MFCD00007241