

N,N-Dimethyl-p-toluidine ≥98.5% (by GC), catalyst grade, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-97-8
:
Formula:C₉H₁₃N
MDL Number:MFCD00008316
:135.21 g/mol
Boiling Pt:211 °C (1013 hPa)
Melting Pt:–15 °C
Density:0.9 g/cm³ (20 °C)
Flash Pt:76 °C
Storage Temperature:Ambient
EINECS:202-805-4
UN:1708
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- APPEARANCE (COLOR):Colorless to Dark Yellow and Colorless to Dark Green-Yellow
- APPEARANCE (FORM):Liquid
- PURITY (GC AREA %):≥ 98.5 %
- DENSITY D20/4:0.930 - 0.945
- REFRACTIVE INDEX N20/D:1.546 - 1.548
- PROTON NMR SPECTRUM:CONFORMS TO STRUCTURE
- RECOMMENDED RETEST PERIOD:10 MONTHS
Specification Test Results
Frequently Bought Together