

N,N-Dimethyl-p-toluidine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-97-8
:
Formula:C₉H₁₃N
MDL Number:MFCD00008316
:135.21 g/mol
Boiling Pt:211 °C (1013 hPa)
Melting Pt:–15 °C
Density:0.9 g/cm³ (20 °C)
Flash Pt:76 °C
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:202-805-4
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.65 g/l (37 °C)
- Melting Point:-15 °C
- Molar Mass:135.21 g/mol
- Boiling Point:215 °C (1013 hPa)
- Vapor Pressure:0.1 hPa (20 °C)
- Flash Point:76 °C
- Refractive Index:1.546 (20 °C, 589 nm)
- Density:0.94 g/cm3 (20 °C)
- Ignition Point:425 °C
- Synonyms:4-Dimethylaminotoluene
- Hill Formula:C9H13N
- Chemical Formula:4-(CH3)C6H4N(CH3)2
Specification Test Results
Related Information
Frequently Bought Together