Order Entry
Czech Republic
ContactUsLinkComponent
3'-Iodoacetophenone, Sigma-Aldrich®
3'-Iodoacetophenone, Sigma-Aldrich®
Catalog # SIAL681164-5G
Supplier:  Merck
CAS Number:  
3'-Iodoacetophenone, Sigma-Aldrich®
Catalog # SIAL681164-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:284 °C (1013 hPa)
Melting Pt:53 °C
Density:1.767 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ100
 :246.05 g/mol
CAS Number:14452-30-3
Formula:C₈H₇IO
MDL Number:MFCD00060283