2-Iodo-1-phenylethane, Sigma-Aldrich®
Catalog # SIAL380490-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:232.06 g/mol
Boiling Pt:125 °C (18 mmHg)
Melting Pt:–2 °C
Density:1.6323 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
CAS Number:17376-04-4
Formula:C₆H₅CH₂CH₂I
MDL Number:MFCD00039408