4-(Trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2991-42-6
:
Formula:C₇H₄ClF₃O₂S
MDL Number:MFCD00042422
:244.62 g/mol
Boiling Pt:49.5…50.5 °C (0.2 mmHg)
Melting Pt:31…33 °C
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ100
Frequently Bought Together