2-(Trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL491624-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:244.62 g/mol
Boiling Pt:123…126 °C (1013 hPa)
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:3261
CAS Number:776-04-5
Formula:C₇H₄ClF₃O₂S
MDL Number:MFCD00051696