
2-(Trifluoromethyl)benzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:776-04-5
:
Formula:C₇H₄ClF₃O₂S
MDL Number:MFCD00051696
:244.62 g/mol
Boiling Pt:133…135 °C (14 mmHg)
Melting Pt:22…24 °C
Density:1.585
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Related Information
Frequently Bought Together