
4-(Trifluoromethoxy)benzylamine, Sigma-Aldrich®
Catalog # SIAL340987-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:191.15 g/mol
Boiling Pt:57 °C (1013 hPa)
Density:1.25 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:300-040-1
:GHS07
CAS Number:93919-56-3
Formula:C₈H₈F₃NO
MDL Number:MFCD00061237