Order Entry
Czech Republic
ContactUsLinkComponent
 
Corrosive
2,4-Difluorobenzylamine, Sigma-Aldrich®
Catalog # SIAL264377-5G
Supplier:  Merck
CAS Number:  
Corrosive
2,4-Difluorobenzylamine, Sigma-Aldrich®
Catalog # SIAL264377-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :143.14 g/mol
Boiling Pt:82…84 °C (15 mmHg)
Density:1.2 g/cm³ (20 °C)
ADR:8,III
 :M-Clarity™ quality level = MQ100
EINECS:276-502-0
UN:2735
 :GHS05
CAS Number:72235-52-0
Formula:C₇H₇F₂N
MDL Number:MFCD00010142