2-Nitro-α,α,α-trifluoro-p-toluic acid, Sigma-Aldrich®
Katalogové číslo SIAL328901-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:235.12 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:320-94-5
Formula:C₈H₄F₃NO₄
MDL Number:MFCD00007142