Order Entry
Czech Republic
ContactUsLinkComponent
4-Fluoro-2-(trifluoromethyl)benzoyl chloride, Sigma-Aldrich®
4-Fluoro-2-(trifluoromethyl)benzoyl chloride, Sigma-Aldrich®
Catalog # SIAL455393-5G
Supplier:  Merck
CAS Number:  
4-Fluoro-2-(trifluoromethyl)benzoyl chloride, Sigma-Aldrich®
Catalog # SIAL455393-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :226.56 g/mol
Boiling Pt:157 °C (1013 hPa)
Density:1.495 g/cm³ (20 °C)
ADR:8,II
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
UN:3265
CAS Number:189807-21-4
Formula:C₈H₃ClF₄O
MDL Number:MFCD00061158