
3'-Fluoroacetophenone, Sigma-Aldrich®
Catalog # SIAL219347-5G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:5 g
- Sekvence:3173
Specifications
About this item
:
Boiling Pt:204 °C (1013 hPa)
Melting Pt:–3 °C
Density:1,12 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:207-245-4
:GHS07
CAS Number:455-36-7
Formula:C₈H₇FO
MDL Number:MFCD00000338