
2',4'-Difluoroacetophenone, Sigma-Aldrich®
Catalog # SIAL264253-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:156.13 g/mol
Boiling Pt:80 °C (1013 hPa)
Density:1.23 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:206-667-6
:GHS07
Storage Temperature:Ambient
CAS Number:364-83-0
Formula:C₈H₆F₂O
MDL Number:MFCD00151261