

α,α,α-Trifluoroacetophenone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:434-45-7
:
Formula:C₈H₅F₃O
MDL Number:MFCD00000420
:174.12 g/mol
Boiling Pt:165…166 °C (760 mmHg)
Melting Pt:–40 °C
Density:1.24 g/cm³
Flash Pt:41 °C
Storage Temperature:Ambient
EINECS:207-103-1
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4580 to 1.4600 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together