Order Entry
Czech Republic
ContactUsLinkComponent
2-Toluoyl chloride 99%
Corrosive
2-Toluoyl chloride 99%
Certificates
undefined
Corrosive
2-Toluoyl chloride 99%

Specification Test Results

  • Appearance:
    Clear colorless to yellow to pink Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5540 to 1.5560 (20°C, 589 nm)
  • Titration RCOCl:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together