2',5'-Dihydroxyacetophenone, Sigma-Aldrich®
Catalog # SIALD107603-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:152.15 g/mol
Boiling Pt:296 °C (1013 hPa)
Melting Pt:199…202 °C
:M-Clarity™ quality level = MQ100
EINECS:207-716-4
CAS Number:490-78-8
Formula:C₈H₈O₃
MDL Number:MFCD00002343