2',5'-Dihydroxyacetophenone for synthesis, Sigma-Aldrich®
Catalog # 8.18284.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:199 - 202 °C
- Molar Mass:152.15 g/mol
- Hill Formula:C8H8O3
- Chemical Formula:2,5-(OH)2-C6H3COCH3
Specification Test Results
About this item
:
:152.15 g/mol
Boiling Pt:296 °C (1013 hPa)
Melting Pt:199…202 °C
:M-Clarity™ quality level = MQ200
EINECS:207-716-4
CAS Number:490-78-8
Formula:C₈H₈O₃
MDL Number:MFCD00002343