
2',6'-Dihydroxyacetophenone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:699-83-2
:
Formula:C₈H₈O₃
MDL Number:MFCD00002270
:152.15 g/mol
Boiling Pt:296 °C (1013 hPa)
Melting Pt:156…159 °C
Storage Temperature:Ambient
EINECS:211-833-6
:GHS07
Specification Test Results
- Appearance:Yellow to light beige Powder
- Infrared spectrum:Conforms
- Melting point:155 to 158 °C
- HPLC:≥98.5 %
- Clarity of solution:Passes test
Specification Test Results
Frequently Bought Together