
2-Hydroxyacetophenone ≥97%
Catalog # 42013.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:5 g
- Sekvence:3173
Specifications
About this item
CAS Number:582-24-1
:
Formula:C₈H₈O₂
Boiling Pt:118…120 °C (11 mmHg)
Melting Pt:86…88 °C
Density:1,0963 g/cm³ (99 °C)
:Packaged under argon.
Storage Temperature:Ambient
EINECS:209-480-8
:GHS07
MDL Number:MFCD00041829