Order Entry
Czech Republic
ContactUsLinkComponent
3',4',5'-Trimethoxyacetophenone, Sigma-Aldrich®
3',4',5'-Trimethoxyacetophenone, Sigma-Aldrich®
Catalog # SIALT68101-25G
Supplier:  Merck
CAS Number:  
3',4',5'-Trimethoxyacetophenone, Sigma-Aldrich®
Catalog # SIALT68101-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :210.23 g/mol
Boiling Pt:318 °C (1013 hPa)
Melting Pt:73…74 °C
 :M-Clarity™ quality level = MQ100
EINECS:214-501-9
CAS Number:1136-86-3
Formula:C₁₁H₁₄O₄
MDL Number:MFCD00008738