3',4',5'-Trimethoxyacetophenone ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1136-86-3
:
Formula:C₁₁H₁₄O₄
MDL Number:MFCD00008738
:210.23 g/mol
Boiling Pt:173…174 °C (10 mmHg)
Melting Pt:79…83 °C
Storage Temperature:Ambient
EINECS:214-501-9
Related Information
Frequently Bought Together