2',4',6'-Trimethoxyacetophenone, Sigma-Aldrich®
Catalog # SIAL630594-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:210.23 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:832-58-6
Formula:C₁₁H₁₄O₄
MDL Number:MFCD00017238