
4'-Methoxyacetophenone ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-06-1
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00008745
:150.18 g/mol
Boiling Pt:258…260 °C
Melting Pt:36…40 °C
Density:1.08
Flash Pt:138 °C (280 °F)
EINECS:202-815-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together