4'-Ethoxyacetophenone
Catalog # PROOMM0474.04
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:164.2 g/mol
Boiling Pt:83 °C (0.04 mmHg)
Boiling Pt:83 °C (0,04 mmHg)
Melting Pt:39 °C
EINECS:216-825-6
CAS Number:1676-63-7
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009095