5-Chloro-2-nitrotoluene, Sigma-Aldrich®
Catalog # SIAL226653-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:171.58 g/mol
Boiling Pt:249 °C (1013 hPa)
Melting Pt:25 °C
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:226-355-3
UN:3457
CAS Number:5367-28-2
Formula:C₇H₆ClNO₂
MDL Number:MFCD00024579