
3,4-Dichlorobenzylamine 98%
Catalog # ACRO113240050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5775 to 1.5795 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
About this item
:
:176.04 g/mol
Boiling Pt:80…82 °C (.6 mmHg)
Density:1.32 g/cm³
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:203-035-1
UN:2735
ADR:8,III
:GHS05
CAS Number:102-49-8
Formula:C₇H₇Cl₂N
MDL Number:MFCD00008114